Anti-NIF3L1 Mouse Monoclonal Antibody [clone: OTI4B5]
ORIGTA503798
: OriGene
- Antibody type:Primary
- Antigen name:NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae)
- Antigen symbol:NIF3L1
- Clonality:Monoclonal
- Clone:OTI4B5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:38.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: NIF3L1 (NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human