Anti-PPARA Mouse Monoclonal Antibody [clone: OTI1E11]
ORIGTA503541
: OriGene
- Antibody type:Primary
- Antigen name:peroxisome proliferator-activated receptor alpha
- Antigen symbol:PPARA
- Clonality:Monoclonal
- Clone:OTI1E11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:145 to 420
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:52 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100
Type: Primary
Antigen: PPARA (peroxisome proliferator-activated receptor alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human