Anti-LGALS3BP Mouse Monoclonal Antibody [clone: OTI2B1]
ORIGTA503459
: OriGene
- Antibody type:Primary
- Antigen name:lectin, galactoside-binding, soluble, 3 binding Protein
- Antigen symbol:LGALS3BP
- Clonality:Monoclonal
- Clone:OTI2B1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:19 to 300
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:65.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: LGALS3BP (lectin, galactoside-binding, soluble, 3 binding protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human