Anti-XPNPEP3 Mouse Monoclonal Antibody [clone: OTI4B8]
ORIGTA503339
: OriGene
- Antibody type:Primary
- Antigen name:X-prolyl aminopeptidase (aminopeptidase P) 3, putative
- Antigen symbol:XPNPEP3
- Clonality:Monoclonal
- Clone:OTI4B8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:56.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: XPNPEP3 (X-prolyl aminopeptidase (aminopeptidase P) 3, putative)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: