Anti-NUDT9 Mouse Monoclonal Antibody [clone: OTI7A12]
ORIGTA503333
: OriGene
- Antibody type:Primary
- Antigen name:nudix (nucleoside diphosphate linked moiety X)-type motif 9
- Antigen symbol:NUDT9
- Clonality:Monoclonal
- Clone:OTI7A12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:38.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: NUDT9 (nudix (nucleoside diphosphate linked moiety X)-type motif 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human