Anti-HSPBP1 Mouse Monoclonal Antibody [clone: OTI1H4]
ORIGTA503317
: OriGene
- Antibody type:Primary
- Antigen name:HSPA (heat shock 70kDa) binding Protein, cytoplasmic cochaperone 1
- Antigen symbol:HSPBP1
- Clonality:Monoclonal
- Clone:OTI1H4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:39.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: HSPBP1 (HSPA (heat shock 70kDa) binding protein, cytoplasmic cochaperone 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human