Anti-EIF4E Mouse Monoclonal Antibody [clone: OTI6C11]
ORIGTA502993
: OriGene
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 4E
- Antigen symbol:EIF4E
- Clonality:Monoclonal
- Clone:OTI6C11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:24.9 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100
Type: Primary
Antigen: EIF4E (eukaryotic translation initiation factor 4E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human