Anti-GRHPR Mouse Monoclonal Antibody [clone: OTI1H8]
ORIGTA502095
: OriGene
- Antibody type:Primary
- Antigen name:Glyoxylate reductase/hydroxypyruvate reductase
- Antigen symbol:GRHPR
- Clonality:Monoclonal
- Clone:OTI1H8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:35.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: GRHPR (glyoxylate reductase/hydroxypyruvate reductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human