Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI5G7]
ORIGTA501558
: OriGene
- Antibody type:Primary
- Antigen name:tripartite motif containing 2
- Antigen symbol:TRIM2
- Clonality:Monoclonal
- Clone:OTI5G7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1 to 100 and 645 to 744
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:81.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: TRIM2 (tripartite motif containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human