Northern Ireland
Order Entry
Anti-GORASP1 Mouse Monoclonal Antibody [clone: OTI5G8]
  ORIGTA501309
 :  OriGene
undefined
Anti-GORASP1 Mouse Monoclonal Antibody [clone: OTI5G8]
  ORIGTA501309
 :  OriGene
 
£582.00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Golgi reassembly stacking protein 1 65kDa
  • Antigen symbol:
    GORASP1
  • Clonality:
    Monoclonal
  • Clone:
    OTI5G8
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    46.3 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

  • Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GORASP1 (golgi reassembly stacking protein 1, 65kDa)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: