Anti-TACC3 Mouse Monoclonal Antibody [clone: OTI1E11]
Catalog # ORIGTA501263
Supplier: OriGene
- Antibody type:Primary
- Antigen name:transforming, acidic coiled-coil containing Protein 3
- Antigen symbol:TACC3
- Clonality:Monoclonal
- Clone:OTI1E11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:90.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
About this item
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: TACC3 (transforming, acidic coiled-coil containing protein 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat