Anti-H6PD Mouse Monoclonal Antibody [clone: OTI2C12]
ORIGTA501213
: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Hexose-6-phosphate dehydrogenase
- Antigen symbol:H6PD
- Clonality:Monoclonal
- Clone:OTI2C12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:88.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: H6PD (hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human