Northern Ireland
Order Entry
Anti-ERMAP Mouse Monoclonal Antibody [clone: OTIC8]
Catalog # ORIGTA500911
Supplier:  OriGene
undefined
Anti-ERMAP Mouse Monoclonal Antibody [clone: OTIC8]
Catalog # ORIGTA500911
Supplier:  OriGene
Price
£582.00
Log in for availability
Unit of Measure 1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    erythroblast membrane-associated Protein (Scianna blood group)
  • Antigen symbol:
    ERMAP
  • Clonality:
    Monoclonal
  • Clone:
    OTIC8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    52.6 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

 

Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:50-1:100

Type: Primary
Antigen: ERMAP (erythroblast membrane-associated protein (Scianna blood group))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human