Anti-CTAG1B Mouse Monoclonal Antibody [clone: OTI3F10]
Catalog # ORIGTA500498
Supplier: OriGene
- Antibody type:Primary
- Antigen name:cancer/testis antigen 1B
- Antigen symbol:CTAG1B
- Clonality:Monoclonal
- Clone:OTI3F10
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:17.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
About this item
Recommended Dilutions: Western Blot: 1:500-1000; Immunofluorescence: 1:50-100
Type: Primary
Antigen: CTAG1B (cancer/testis antigen 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human