Anti-PMEL Mouse Monoclonal Antibody [clone: OTI10E5]
ORIGTA500424
: OriGene
Specifications
- Antibody type:Primary
- Antigen name:premelanosome Protein
- Antigen symbol:PMEL
- Clonality:Monoclonal
- Clone:OTI10E5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:70.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50 Immunofluorescence: 1:100 Flow Cytometry: 1:500
Type: Primary
Antigen: PMEL (premelanosome protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: