Anti-SSB Mouse Monoclonal Antibody [clone: OTI3H9]
ORIGTA500410
: OriGene
- Antibody type:Primary
- Antigen name:Sjogren syndrome antigen B (autoantigen La)
- Antigen symbol:SSB
- Clonality:Monoclonal
- Clone:OTI3H9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:46.8 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
About this item
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l
Type: Primary
Antigen: SSB (Sjogren syndrome antigen B (autoantigen La))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: