Anti-CHEK2 Mouse Monoclonal Antibody [clone: OTI2H5]
ORIGTA500384
: OriGene
- Antibody type:Primary
- Antigen name:Checkpoint Kinase 2
- Antigen symbol:CHEK2
- Clonality:Monoclonal
- Clone:OTI2H5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:60.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
- Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Flow Cytometry: 1:100; Immunoprecipitation: 2μg/500μl
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: