Anti-IL3 Mouse Monoclonal Antibody [clone: OTI3B11]
ORIGTA500182
: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 3
- Antigen symbol:IL3
- Clonality:Monoclonal
- Clone:OTI3B11
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:20 to 152
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:15.0 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100
Type: Primary
Antigen: IL3 (interleukin 3 (colony-stimulating factor, multiple))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human