Northern Ireland
Order Entry
Anti-DDK Tag Monoclonal Antibody [clone: OTI4C5]
Anti-DDK Tag Monoclonal Antibody [clone: OTI4C5]
  ORIGTA50011-100
 :  OriGene
Anti-DDK Tag Monoclonal Antibody [clone: OTI4C5]
  ORIGTA50011-100
 :  OriGene
 
£536.00
   
  1 * 100 µl

 

  • Antibody type:
    Tag
  • Antigen name:
    DDK Tag
  • Antigen symbol:
    DDK
  • Clonality:
    Monoclonal
  • Clone:
    OTI4C5
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide
  • Sequence:
    dykddddk
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:2000; Immunoprecipitation: 1:200; Immunofluorescence: 1:1000; Flow Cytometry: 1:1000

Type: Tag
Antigen: DDK Tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity: