Northern Ireland
Order Entry
Anti-MAPK8 Mouse Monoclonal Antibody [clone: OTI4E3]
  ORIGTA500081
 :  OriGene
undefined
Anti-MAPK8 Mouse Monoclonal Antibody [clone: OTI4E3]
Catalog # ORIGTA500081
Supplier:  OriGene
Price
£582.00
Log in for availability
Unit of Measure 1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase 8
  • Antigen symbol:
    MAPK8
  • Clonality:
    Monoclonal
  • Clone:
    OTI4E3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    1 to 384
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    44.0 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

 

 

Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: MAPK8 (mitogen-activated protein kinase 8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human