Northern Ireland
Order Entry
Anti-GSC Mouse Monoclonal Antibody [clone: OTI1D7]
  ORIGTA500023
 :  OriGene
undefined
Anti-GSC Mouse Monoclonal Antibody [clone: OTI1D7]
  ORIGTA500023
 :  OriGene
 
£582.00
   
  1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Goosecoid homeobox
  • Antigen symbol:
    GSC
  • Clonality:
    Monoclonal
  • Clone:
    OTI1D7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Amino acid number:
    107 to 257
  • Storage buffer:
    PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    28.0 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

 

Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: GSC (goosecoid homeobox)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human