Anti-GSK3B Rabbit Polyclonal Antibody
ORIGTA317698
: OriGene
- Antibody type:Primary
- Antigen name:Glycogen synthase kinase 3 beta
- Antigen symbol:GSK3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Modification:phosphoSer9
- Storage buffer:0,02 M potassium phosphate, 0,15 M sodium chloride, pH 7,2, 0,01% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
Recommended Dilutions: ELISA: 1:10000-1:50000; Immunofluorescence: 1:200-1:1000; Immunochemistry: 5 µg/ml; Immunoprecipitation; Western Blot: 1:500-1:3000
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human