Anti-FFAR4 Rabbit Polyclonal Antibody
ORIGTA317593
: OriGene
- Antibody type:Primary
- Antigen name:Free fatty acid receptor 4
- Antigen symbol:FFAR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:N-terminal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
- Recommended Dilutions: Immunochemistry: 1 µg/ml
Type: Primary
Antigen: FFAR4 (free fatty acid receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human