Anti-SLC5A9 Rabbit Polyclonal Antibody
Catalog # ORIGTA317343
Supplier: OriGene
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/sugar cotransporter), member 9
- Antigen symbol:Solute carrier family 5, member 9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Epitope:C-terminal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Pk:50 µG
- Recommended Dilutions: Immunochemistry: 10 µg/ml
Type: Primary
Antigen: SLC5A9 (solute carrier family 5 (sodium/sugar cotransporter), member 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human