Northern Ireland
Order Entry
Anti-CDK1 Mouse Monoclonal Antibody [clone: 26E11]
  ORIGTA316617
 :  OriGene
Anti-CDK1 Mouse Monoclonal Antibody [clone: 26E11]
Catalog # ORIGTA316617
Supplier:  OriGene
Price
£650.00
Log in for availability
Unit of Measure 1 * 50 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent kinase 1
  • Antigen symbol:
    CDK1
  • Clonality:
    Monoclonal
  • Clone:
    26E11
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulfate precipitation
  • Pk:
    50 µl

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:200 Western Blot: 1:1000

Type: Primary
Antigen: CDK1 (cyclin-dependent kinase 1)
Clonality: Monoclonal
Clone: 26E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat