Northern Ireland
Order Entry
Anti-ZMYND11 Rabbit Polyclonal Antibody
Anti-ZMYND11 Rabbit Polyclonal Antibody
Catalog # ORIGTA315102
Supplier:  OriGene
Anti-ZMYND11 Rabbit Polyclonal Antibody
Catalog # ORIGTA315102
Supplier:  OriGene
Price
£495.00
Log in for availability
Unit of Measure 1 * 100 µl

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    zinc finger, MYND-type containing 11
  • Antigen symbol:
    ZMYND11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:5000

Type: Primary
Antigen: ZMYND11 (zinc finger, MYND-type containing 11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat