Anti-POLE Rabbit Polyclonal Antibody
Catalog # ORIGTA314359
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (DNA directed), epsilon, catalytic subunit
- Antigen symbol:POLE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:50-1:100 ELISA: 1:40000
Type: Primary
Antigen: POLE (polymerase (DNA directed), epsilon, catalytic subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse