Anti-SLC27A4 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP43901_P050EA 339 GBP
AVIVARP43901_P050
Anti-SLC27A4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC27A4 (solute carrier family 27 (fatty acid transporter), member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR