Anti-VSX1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP39108_T100EA 303 GBP
AVIVARP39108_T100
Anti-VSX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VSX1 (visual system homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR