Anti-Rhox8 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
AVIVARP37849_P050EA 348 GBP
AVIVARP37849_P050
Anti-Rhox8 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Rhox8
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR