Northern Ireland
Order Entry
Anti-COPS2 Rabbit Polyclonal Antibody
Certificates
Anti-COPS2 Rabbit Polyclonal Antibody

 

  • Catalog No:
  • AVIVARP34189_P050
  • AVIVARP34189_T100
  • Antigen symbol:
  • COPS2
  • COPS2
  • Antigen name:
  • COP9 signalosome subunit 2
  • COP9 signalosome subunit 2
  • Antigen synonyms:
  • ALIEN|SGN2|CSN2|TRIP15
  • ALIEN|SGN2|CSN2|TRIP15
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 9318
  • 9318
  • Western blot:
  • Yes
  • Yes
  • Epitope:
  • N-terminal
  • N-terminal
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Related Information

Product Information

  •  
     
    Pk
     
     
  • AVIVARP34189_P050
    ARP34189_P050
    50 µG
       
       
    1 * 50 µG
    1 * 50 µG - £353.00
    £353.00
    1 * 50 µG
       £353.00

     
     
    Specifications
    Antigen symbol
    COPS2
    Antigen name
    COP9 signalosome subunit 2
    Environmentally Preferable
    Antigen synonyms
    ALIEN|SGN2|CSN2|TRIP15
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Gene ID
    9318
    Isotype
    Western blot
    Yes
    Epitope
    N-terminal
    Cross adsorption
    none
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • AVIVARP34189_T100
    ARP34189_T100
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG - £307.00
    £307.00
    1 * 100 µG
       £307.00

     
     
    Specifications
    Antigen symbol
    COPS2
    Antigen name
    COP9 signalosome subunit 2
    Environmentally Preferable
    Antigen synonyms
    ALIEN|SGN2|CSN2|TRIP15
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Gene ID
    9318
    Isotype
    Western blot
    Yes
    Epitope
    N-terminal
    Cross adsorption
    none
    Storage temperature
    Cool
    Shipping temperature
    Ice