Anti-MKL1 Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-6973REA 313 GBP
BOSSBS-6973R
Anti-MKL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MKL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR