Anti-CD69 Rabbit Polyclonal Antibody
Catalog # BOSSBS-2499R
: Bioss
- Antibody type:Primary
- Antigen name:CD69 molecule
- Antigen symbol:CD69
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:EA1|MLR-3|AIM|GP32/28|CLEC2C|BL-AC/P26
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Type: Primary
Antigen: CD69
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: