Anti-PSCA Rabbit Polyclonal Antibody

Supplier: Bioss
BOSSBS-0669REA 672 GBP
BOSSBS-0669R
Anti-PSCA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PSCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR