Large cryogenic containers, with narrow neck, SD series
478-0650EA 35540 GBP
VWRI478-0650 VWRI478-0651
Large cryogenic containers, with narrow neck, SD series
The SD Series are designed for long-term storage at extreme low and stable temperatures. The cryogenic containers are designed with high efficiency multi-layer super-insulation, narrow neck opening to limit nitrogen evaporation and temperature fluctuations, providing a reliable and efficient cryopreservation for samples collected in straws, cryovials or blood bags.
  • Narrow neck opening of the container offers lowest possible evaporation rate and extreme low storage temperatures, even in gas phase
  • Turning table with rotating handle on the outside of the vessel
  • Gas spring assisted opening lid with locking device (optional)
  • Possibility of storing in liquid, gas or dry phase
  • Wide range of polycarbonate, aluminium and stainless steel racks available
The unique turning system of the inner basket with a turning handle positioned on the outside, ensures that the user does not need to insert his/her hands in the cryovessel.
Certifications: The vessels are made in France with stainless steel, according to the highest quality levels.
Compliant with European Medical Device Directive 93/42/EEC.