Diethyl ether stabilised USP, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Ether
, 2,2'-Oxybisethane
, Ethoxyethane
, Ethyl Ether
, 2,2'-Oxydiethane
Stabilisation:
Stabilised with BHT (2,6-Di-tert-butyl-p-cresol, 2,6-Di-tert-butyl-4-methylphenol, Ionol) and ethanol
9239-22EA
114
GBP
9239-22
9239-R
9239-07
Diethyl ether stabilised USP, J.T.Baker®
Diethyl ether
Formula:
(CH₃CH₂)₂O MW: 74,12 g/mol Boiling Pt: 35 °C (1013 hPa) Melting Pt: –123 °C Density: 0,7135 g/cm³ (20 °C) Flash Pt: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Specific Gravity at 25°/25°C | 0.713 - 0.716 |
USP - Acidity | Passes Test |
USP - Nonvolatile Residue | ≤0.003 % |
USP - Aldehydes | Passes Test |
USP - Peroxide (as H₂O₂) | ≤0.3 ppm |
USP - Low Boiling Hydrocarbons | ≤0.2 % |
USP - Preservative (C₂H₅OH), For Information Only | |
USP - Preservative (H₂O), (w/w) | ≤0.5 % |
USP - Preservative (BHT) | ≥7 ppm |
Only the Class 3 solvent Ethanol is likely to be present as a preservative. The actual concentration is reported. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...