Hybridisation incubator, 7601
Supplier: GFL
390-8500EA 0 GBP
390-8500
Hybridisation incubator, 7601
The 7601 hybridisation incubator is a specialist for individual cases. The small quantity of hybridisation liquid that is required enables the user to work with high concentrations of probes or antibodies. Other applications are also possible. Instead of the rotating rack, the incubator’s interior can be equipped with up to five non-tilt and pull-out trays (optional accessories). The rotating rack to accept the hybridisation vessels is driven by a geared motor via a sliding clutch. It is easily removable and can be loaded outside the incubator.
  • Thermostatic over-temperature cutout to protect the heating element
  • High resolutions of the detections and even results due to the constancy of 10 rpm
  • Soft-touch keys with clear symbols
  • Protection against unintended alterations by two-finger-operation
The ventilator ensures optimum temperature distribution throughout the cabinet interior. Electronical monitoring of the temperature controller triggers a visual alarm in case of a fault, i.e. if the set value is exceeded by 4 ºC. The heating is switched off, the cause of the fault is shown on the LED display.
Clear view of the interior cabinet through a large 16 mm thick heat insulating acrylic glass window is possible. The interior parts of the unit, such as cabinet walls, air baffle plate, interior door frame, rotating rack and the removable drip tray under the rotating rack are made of stainless steel, the robust housing is made of electrolytically galvanised sheet steel.
Delivery information: Supplied with two clip wheels, four bottles (Ø 51 mm) and eight spring clips (Ø 51 mm).