ESD safety ankle boots, lace-up, Static Control, 31475
Supplier: Abeba
Static Control
ABEB31475/45EA 150 GBP
ABEB31475/45 ABEB31475/46 ABEB31475/43 ABEB31475/44 ABEB31475/38 ABEB31475/39 ABEB31475/47 ABEB31475/36 ABEB31475/37 ABEB31475/41 ABEB31475/42 ABEB31475/40
ESD safety ankle boots, lace-up, Static Control, 31475
Metal-free ESD ankle boot made from smooth leather, reflective material, and a nitrile sole resistant to acids, bases (cleaning agents) and fuel.
  • ASC (anti-slip contact) nitrile rubber sole: Three different rubber compounds and patterns ensure best slip resistance and protection
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Metal-free, puncture resistant midsole
  • Composite toecap (200 J)
Colour: Black
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20347:2012; S3, SRA