Cryostats, CryoStar™ NX70
Supplier: Epredia
CryoStar™
720-2714EA 0 GBP
720-2714 720-2712 720-2713
Cryostats, CryoStar™ NX70
Microtomes and Microtome Assemblies Microtomes
The CryoStar NX70 can accomodate the needs of both research and clinical laboratories.
  • Form-fitting design, integrated height control and motorised sectioning
  • Rapid Response Temperature Control adjusts quickly to settings changes and maintains temperature through periods of high-volume use
  • Motorised cutting reduces user fatigue and avoids repetitive motion injury
  • Independent blade holder and object cooling keeps cutting area at set temperatures even during periods of high use
  • Cross-roller bearing design provides years of maintenance-free operation