You Searched For: Microbiology Learning Activities


10  results were found

SearchResultCount:"10"

Sort Results

List View Easy View

Rate These Search Results

Catalog Number: (EDVO221)
Supplier: EDVOTEK EUROPE LTD
Description: In this experiment, students see a blue colour change in transformed cells due to the switching on of a gene. The pGAL plasmid gives a blue colour dye...
UOM: 1 * 1 items


Catalog Number: (EDVO223/AP08)
Supplier: EDVOTEK EUROPE LTD
Description: In this experiment, transformed cells take up a plasmid containing the Green Fluorescent Protein (GFP) gene. The GFP gene was isolated from the jellyf...
UOM: 1 * 1 items


Catalog Number: (EDVO255)
Supplier: EDVOTEK EUROPE LTD
Description: When bacteria are used to make medicinally useful proteins by transformation, the protein of interest must be separated from all of the other cellular...
UOM: 1 * 1 KIT


Catalog Number: (EDVO222)
Supplier: EDVOTEK EUROPE LTD
Description: The Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria is used extensively in all areas of science. Many organisms have been ...
UOM: 1 * 1 items

MSDS


Catalog Number: (BE-301)
Supplier: G-Biosciences
Description: Bacterial conjugation is a naturally occurring process that allows the transfer of DNA from one bacterium to another, which allows the transfer of gen...
UOM: 1 * 1 KIT


Catalog Number: (BE-202)
Supplier: G-Biosciences
Description: The Gram staining method was first described in 1844 by the Danish bacteriologist Hans Christian Gram, after whom the test was named. The Gram staini...
UOM: 1 * 1 KIT


Catalog Number: (BE-201)
Supplier: G-Biosciences
Description: This kit teaches aseptic handling techniques and cultivation of bacteria in liquid culture media and on solid phase agar plates. This kit is designed ...
UOM: 1 * 1 KIT


Catalog Number: (BE-203)
Supplier: G-Biosciences
Description: Antibiotics play a crucial role in the manipulation, screening and killing of bacteria in a range of biotechnology processes. This kit specifically t...
UOM: 1 * 1 KIT


Catalog Number: (EDVO615)
Supplier: EDVOTEK EUROPE LTD
Description: Educational Materials, Microbiology, Application: Microbiology
UOM: 1 * 1 mL


Catalog Number: (EDVO616)
Supplier: EDVOTEK EUROPE LTD
Description: Educational Materials, Microbiology, Application: Microbiology, ReadyPourâ„¢ LB agar with ampicillin
UOM: 1 * 170 mL


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 10 of 10
no targeter for Bottom