Designed with durability and comfort in mind, the GasAlert Extreme reliably monitors for single gas hazard within its wide range of available toxic gas models.
- Direct, real-time readings on large LCD
- Water resistant with easy on/off operation
- Full function self-test of sensor, battery status, circuit integrity and audible/visual alarms on start up
- Bright wide angled visual alarm bars
- Built-in concussion-proof boot
This single gas detector offers extended longevity with a two-year field-replaceable battery and sensor.
Calibration is a simple automatic procedure and is compatible with MicroDock II automatic test and calibration system.
A range of user options, multi-language display and datalogging functionality make the GasAlert Extreme a solution for many applications.
Certifications: EMC Directive 2004/108/EC; IP 66/67. IECEx: Ex ia IIC T4.
Delivery information: Supplied complete with specified sensor, stainless steel alligator clip and concussion-proof housing, calibration/test cap and hose, manual and multi-language CD manual.