Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
/
Northern Ireland
Products
Services
Digital Solutions
Applications
Support
About
£0.00
Order Entry
Home
Methyl 4-amino-3-bromo-5-nitrobenzoate
Methyl-4-amino-3-bromo-5-nitrobenzoate 97%
Methyl-4-amino-3-bromo-5-nitrobenzoate 97%
Methyl-4-amino-3-bromo-5-nitrobenzoate 97%
H33216.06
:
Thermo Fisher Scientific
:
Methyl-4-amino-3-bromo-5-nitrobenzoate 97%
H33216.06
:
Thermo Fisher Scientific
:
£180.00
1 * 5 g
-
+
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
275.06 g/mol
:
107…109 °C
:
GHS07
:
Ambient
:
105655-17-2
:
C₈H₇BrN₂O₄
:
MFCD08443938