Mops, M-Zone™ MegaSorb™
Supplier: Micronova
MegaSorb™
MICRMZCRMS-10IREA 866 GBP
MICRMZCRMS-10IR
Mops, M-Zone™ MegaSorb™
Mops
This innovative, low-linting, and highly absorbent microfiber mop head features microfiber tubes which draw up and trap small particles more effectively than standard poly/cotton mops which roll over the dirt. Ideal for laying down disinfectant and drawing up spills.
  • Can be laundered and re-used, and will hold up to alcohol, strong disinfectants, and steam sterilization