1-(Trimethylsilyl)imidazole for gas chromatography, Supelco®

Supplier: Merck

Synonyms: TSIM1-(Trimethylsilyl)-1H-imidazoleTMSIN-(Trimethylsilyl)imidazoleN-(Trimethylsilyl)-1H-imidazole

M-Clarity™ quality level = MQ100

1.09771.0005EA 183 GBP
1.09771.0005
1-(Trimethylsilyl)imidazole for gas chromatography, Supelco®
1-(Trimethylsilyl)imidazole
Formula: C₆H₁₂N₂Si
MW: 140,26 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0,95 g/cm³ (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00005280
CAS Number: 18156-74-6
EINECS: 242-040-3

Order Now

Specification Test Results

Solubility (20 °C) Risk of violent reaction.
Melting Point -42 °C
Molar Mass 140.25 g/mol
Boiling Point 212 °C (decomposition)
Flash Point 61 °C DIN 51755 Part 1
Density 0.95 g/cm3 (25 °C)
Ignition Point 540 °C DIN 51794
Viscosity kinematic 2.5 mm2/s (25 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR