Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Northern Ireland
Order Entry
Order Status
Your Account
Cart
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
£0.00
Order Entry
Home
L(+)-Lyxose
L(+)-Lyxose
L(+)-Lyxose
L(+)-Lyxose
MOLEM26791145
:
Molekula
:
L(+)-Lyxose
Catalog # MOLEM26791145
Supplier:
Molekula
CAS Number:
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 1 g
-
+
Inquire For Price
Specifications
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Specifications
About this item
Product Details
:
:
150.13 g/mol
:
Ambient
:
217-763-2
:
1949-78-6
:
C₅H₁₀O₅
:
MFCD00064111
Product Details