Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

belts

https://ni.vwr.com/stibo/smallweb/std.lang.all/49/34/18944934.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"15"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Portwest Clothing
Description:   Glove clip in acetal copolymer, for attachment to belt loops.
Supplier:  Portwest Clothing
Description:   Hardwearing two-ply PVC belt finished with a heavy-duty metal buckle. Can be worn with any trouser.
Supplier:  FRISTADS KANSAS GROUP
Description:   This belt made of 100% cotton (canvas) features a metal buckle.
Catalog Number: (KWIN100075940)
Supplier:  FRISTADS KANSAS GROUP
Description:   Tool belt made of 100% cotton for handyworkers, carpenters etc.
UOM:  1 * 1 items
Supplier:  FRISTADS KANSAS GROUP
Description:   These ergonomically designed leather belts distribute the weight of your tools more evenly and form the base of the Ergo carrying system to relieve hi...
Supplier:  FRISTADS KANSAS GROUP
Description:   Soft, ergonomic belts made of polyester/PE foam for improved comfort. With 70 mm padded polyester tube and 40 mm nylon strap with metal buckle.
Supplier:  FRISTADS KANSAS GROUP
Description:   This belt made of 100% polyester features a soft buckle.
Catalog Number: (113-6766)
Supplier:  FRISTADS KANSAS GROUP
Description:   Leather belt with antique silver buckle.
UOM:  1 * 1 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 15
  1  2  Next