Flow indicator
Supplier: SciLabware
AZLOFWY053EA 97 GBP
AZLOFWY053
Flow indicator
Maximum operating temperature: 60 °C
  • Indicates flow of liquid or gas
  • Transparent robust PMP moulding with coloured paddle wheel
  • Operates at low flow rates - the impeller starts to turn at approximately 150 ml/minute
Caution: Not for use with some organic solvents, strong acids or strong bases. Please check compatibility of flow materials with PMP at relevant temperature(s).