E.Z.N.A.® Yeast DNA Kit
Supplier: Omega Bio-Tek
D3370-01EA 88.5 GBP
OMEGD3370-01
E.Z.N.A.® Yeast DNA Kit
The E.Z.N.A.® Yeast DNA Kit is comprised of a quick and efficient lysis procedure to isolate of high quality genomic, YAC, or plasmid DNA from yeast. There are no organic extractions, thus reducing plastic waste and hands-on time to allow for reliable isolation of total cellular DNA from a wide variety of yeast species. Purified DNA is suitable for PCR, restriction enzyme digestion, and hybridisation techniques to be processed in parallel.
  • Reproducible DNA purification from variety of sample sources
  • High yield - glass beads and enzymatic digestion for cell lysis