Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 1:00 am CET Saturday through 0:30 am Monday. Orders placed will be processed after maintenance is complete.

Nitrogen generator, Genius 3010, Peak Scientific

Supplier: PEAK SCIENTIFIC INSTRUMENTS LT
564-0119EA 0 GBP InStock
564-0119
Nitrogen generator, Genius 3010, Peak Scientific
Gas Generators Nitrogen Generators
The Genius 3010 nitrogen generator is designed specifically as a stand-alone system to provide gas to single LC-MS applications which require a high nitrogen flow.

  • No external compressor required
  • Insulated compressor compartment for minimum disruption

Max. outlet gas pressure: 6,90 bar

Min./Max. operating temperature: 5 to 35 °C

Max. relative humidity: 80% non condensing

Gas outlets: 1× 1/4" BSPP

Delivery information: Supplied with caster wheels for easy mobility.

Order Now
 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty