Northern Ireland
Order Entry
Anti-GST Antibody (HRP (Horseradish Peroxidase))
Catalog # RPN1236
Supplier:  Cytiva
undefined
Anti-GST Antibody (HRP (Horseradish Peroxidase))
  RPN1236
 :  Cytiva
Price
£392.00
Log in for availability
Unit of Measure 1 * 75 µl

Specifications

  • Antigen name:
    Glutathione-S-Transferase
  • Antigen symbol:
    GST
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Pk:
    75 µl

Specifications

About this item